missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VAPB Protein
A cDNA sequence encoding the VAPB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13922-20ug
Additional Details : Weight : 0.01000kg
Specifications
VAPB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAKVEQVLSL EPQHELKFRG PFTDVVTTNL KLGNPTDRNV CFKVKTTAPR RYCVRPNSGI IDAGASINVS VMLQPFDYDP NEKSKHKFMV QSMFAPTDTS DMEAVWKEAK PEDLMDSKLR CVFELPAEND KPHDVEINKI ISTTASKTET PIVSKSLSSS LDDTEVKKVM EECKRLQGEV QRLREENKQF KEEDGLRMRK TVQSNSPISA LAPTGKEEGL ST | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The VAPB solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |