missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human VAMP3/Cellubrevin Protein

Product Code. 18273655 Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.50mg
This item is not returnable. View return policy

Product Code. 18273655

Brand: Novus Biologicals™ NBC1183460.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-77 of Human VAMP3/Cellubrevin The Recombinant Human VAMP3/Cellubrevin Protein is derived from E. coli. The Recombinant Human VAMP3/Cellubrevin Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/ml
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl pH 7.5, 10% glycerol
Gene ID (Entrez) 9341
Molecular Weight (g/mol) 8.7 kDa
Quantity 0.5 mg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1 microgram of protein (determined by LAL method)
Gene Symbol VAMP3
Cross Reactivity VAMP3/Cellubrevin
Species Human
Purity or Quality Grade >95% pure by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.