missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VAMP1 Protein
A cDNA sequence encoding the VAMP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13915-1mg
Additional Details : Weight : 0.01000kg
Specifications
VAMP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSAPAQPPAE GTEGTAPGGG PPGPPPNMTS NRRLQQTQAQVEEVVDIIRV NVDKVLERDQ KLSELDDRAD ALQAGASQFE SSAAKLKRKY W | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein solution (1 mg/ml) contains 1X PBS and 1mM EDTA. |