missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human V-type proton ATPase subunit d 1 Protein
A DNA sequence encoding the Homo sapiens (Human) V-type proton ATPase subunit d 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5699-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004682.2 | |
V-type proton ATPase subunit d 1 Protein | |
Research Use Only | |
ATP6V0D1 | |
Recombinant Protein | |
E. coli | |
MSFFPELYFN VDNGYLEGLV RGLKAGVLSQ ADYLNLVQCE TLEDLKLHLQ STDYGNFLAN EASPLTVSVI DDRLKEKMVV EFRHMRNHAY EPLASFLDFI TYSYMIDNVI LLITGTLHQR SIAELVPKCH PLGSFEQMEA VNIAQTPAEL YNAILVDTPL AAFFQDCISE QDLDEMNIEI IRNTLYKAYL ESFYKFCTLL GGTTADAMCP ILEFEADRRA FIITINSFGT ELSKEDRAKL FPHCGRLYPE GLAQLARADD YEQVKNVADY YPEYKLLFEG AGSNPGDKTL EDRFFEHEVK LNKLAFLNQF HFGVFYAFVK LKEQECRNIV WIAECIAQRH RAKIDNYIPI F | |
Greater than 80% as determined by SDS-PAGE. |
9114 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |