missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UV excision repair protein RAD23 homolog A Protein

A DNA sequence encoding the Homo sapiens (Human) UV excision repair protein RAD23 homolog A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6578-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004300

  • £2010.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001257291.1
UV excision repair protein RAD23 homolog A Protein
Research Use Only
RAD23A
Recombinant Protein
Yeast
MAVTITLKTL QQQTFKIRME PDETVKVLKE KIEAEKGRDA FPVAGQKLIY AGKILSDDVP IRDYRIDEKN FVVVMVTKTK AGQGTSAPPE ASPTAAPESS TSFPPAPTSG MSHPPPAARE DKSPSEESAP TTSPESVSGS VPSSGSSGRE EDAASTLVTG SEYETMLTEI MSMGYERERV VAALRASYNN PHRAVEYLLT GIPGSPEPEH GSVQESQVSE QPATEAAGEN PLEFLRDQPQ FQNMRQVIQQ NPALLPALLQ QLGQENPQLL QQISRHQEQF IQMLNEPPGE LADISDVEGE VGAIGEEAPQ MNYIQVTPQE KEAIERLKAL GFPESLVIQA YFACEKNENL AANFLLSQNF DDE
Greater than 80% as determined by SDS-PAGE.
5886
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers