missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UTI Protein

A cDNA sequence encoding the UTI was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10909-2.5mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955538

  • £99.15 / 2.50mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

UTI Protein
Research Use Only
Human UTI has an activity of 2390IU/mg.
Human
Untagged
2.5 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Human
Lyophilized from a (1 mg/ml) solution containing no additives.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers