missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human USP2 Catalytic Domain Protein

Product Code. 18371708
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371708 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18371708 Supplier R&D Systems™ Supplier No. E504050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Recombinant Human USP2 Catalytic Domain is a Ubiquitin-specific deconjugating enzyme. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human USP2 Catalytic Domain concentration of 1-5 nM.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75604
For Use With (Application) Enzyme Activity
Formulation 2.0 mg/mL (50 μM) in 50 mM HEPES pH 8.0, 150 mM NaCl, 0.1 mM EDTA, and 1 mM DTT.
Gene ID (Entrez) 9099
Molecular Weight (g/mol) M.W. (theoretical): 40 kDa
Name USP2 Catalytic Domain
Quantity 50 μg
Source E. coli-derived human USP2 protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Symbol USP2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Protein USP2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.