missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UQCRC1 Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | UQCRC1 Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSEFTATFA QALQFVPETQ VSLLDNGLRV ASEQSSQPTC TVGVWIDVGS RFETEKNNGA GYFLEHLAFK GTKNRPGSAL EKEVESMGAH LNAYSTREHT AYYIKALSKD LPKAVELLGD IVQNCSLEDS QIEKERDVIL REMQENDASM RDVVFNYLHA TAFQGTPLAQ AVEGPSENVR KLSRADLTEY LSTHYKAPRM VLAAAGGVEH QQLLDLAQKH LGGIPWTYAE DAVPTLTPCR FTGSEIRHRD DALPFAHVAI AVEGPGWASP DNVALQVANA IIGHYDCTYG GGVHLSSPLA SGAVANKLCQ SFQTFSICYA ETGLLGAHFV CDRMKIDDMM FVLQGQWMRL CTSATESEVA RGKNILRNAL VSHLDGTTPV CEDIGRSLLT YGRRIPLAEW ESRIAEVDAS VVREICSKYI YDQCPAVAGY GPIEQLPDYN RIRSGMFWLR F |
| Buffer | UQCRC1 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction