missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UPRT Protein
A cDNA sequence encoding the UPRT was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13902-10ug
This item is not returnable.
View return policy
Specifications
UPRT Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMATELQC PDSMPCHNQQ VNSASTPSPE QLRPGDLILD HAGGNRASRA KVILLTGYAH SSLPAELDSG ACGGSSLNSE GNSGSGDSSS YDAPAGNSFL EDCELSRQIG AQLKLLPMND QIRELQTIIR DKTASRGDFM FSADRLIRLV VEEGLNQLPY KECMVTTPTG YKYEGVKFEK GNCGVSIMRS GEAMEQGLRD CCRSIRIGKI LIQSDEETQR AKVYYAKFPP DIYRRKVLLM YPILSTGNTV IEAVKVLIEH GVQPSVIILL SLFSTPHGAK SIIQEFPEIT ILTTEVHPVA PTHFGQKYFG TD | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
UPRT protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |