missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human UCK His Protein

Product Code. p-7104604 Shop All Bio Techne Products
missing translation for 'orderingAttributeHoverText'
Quantity:
0.1 mg
0.5 mg
missing translation for 'unitSize'
0.1mg
0.5mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 18106858

missing translation for 'mfr': Novus Biologicals™ NBP2517050.1MG

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 143-273 of Human UCK The Recombinant Human UCK Protein is derived from E. coli. The Recombinant Human UCK Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Spécification

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0) containing 10% glycerol 0.4M UREA
Gene ID (Entrez) 83549
Molecular Weight (g/mol) TMW: 17.5kDa
Quantity 0.1 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias Cytidine monophosphokinase 1, EC 2.7.1.48, FLJ12255, Uridine monophosphokinase 1, uridine-cytidine kinase 1, URK1UCK 1
Common Name UCK
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >80%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Afficher plus Afficher moins

For research use only and is not approved for use in humans or in clinical diagnosis

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.