missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ubiquitin-like protein FUBI Protein

A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-like protein FUBI, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8387-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004361

  • £1735.00 / 1mg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001988.1
Ubiquitin-like protein FUBI Protein
Research Use Only
FAU
Recombinant Protein
E. coli
MQLFVRAQEL HTFEVTGQET VAQIKAHVAS LEGIAPEDQV VLLAGAPLED EATLGQCGVE ALTTLEVAGR MLGG
Greater than 90% as determined by SDS-PAGE.
2197
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers