missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human UBE2R1/CDC34 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1000 μg
5 μg
500 μg
Unit Size:
1000µg
100µg
500µg
5µg
Description
Data is not available.
Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | A 0.2 μm filtered concentrated solution in 50 mM HEPES, pH 7.0, 125 mM NaCl, 10 % Glycerol, 1 mM DTT. |
| Gene ID (Entrez) | 997 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 26.7 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C. Avoid freeze/thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of UBE2R1/CDC34 as determined by LAL method. |
| Gene Alias | cell division cycle 34, cell division cycle 34 homolog (S. cerevisiae), E2-CDC34, EC 6.3.2.19, UBC3, UBE2R1UBCH3, ubiquitin carrier protein, ubiquitin-conjugating enzyme E2 R1, Ubiquitin-conjugating enzyme E2-32 kDa complementing, Ubiquitin-conjugating enzyme E2-CDC34, Ubiquitin-protein ligase R1 |
| Gene Symbol | CDC34 |
| Research Category | Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction