missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human UBE2K/E2-25K His (N-Term) Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
Specifications
Specifications
| Accession Number | P61086 |
| For Use With (Application) | PAGE |
| Formulation | A 0.2 um filtered concentrated solution in 50 mM HEPES, pH 8.0, with 100 mM NaCl, 10 % Glycerol, 1 mM DTT. |
| Gene ID (Entrez) | 3093 |
| Molecular Weight (g/mol) | 23.4 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20C. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of UBE2K/E2-25K as determined by LAL method. |
| Gene Alias | E2(25K), E2-25K, EC 6.3.2.19, HIP-2, huntingtin interacting protein 2, Huntingtin-interacting protein 2, HYPG, ubiquitin carrier protein, ubiquitin-conjugating enzyme E2 K, Ubiquitin-conjugating enzyme E2(25K), Ubiquitin-conjugating enzyme E2-25 kDa, Ubiquitin-conjugating enzyme E2-25K, ubiquitin-conjugating enzyme E2K (UBC1 homolog, yeast), ubiquitin-protein ligase |
| Gene Symbol | UBE2K |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction