missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UBE2G Protein

A cDNA sequence encoding the UBE2G was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13859-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954929

  • £2975.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

UBE2G Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMTELQSA LLLRRQLAEL NKNPVEGFSA GLIDDNDLYR WEVLIIGPPD TLYEGGVFKA HLTFPKDYPL RPPKMKFITE IWHPNVDKNG DVCISILHEP GEDKYGYEKP EERWLPIHTV ETIMISVISM LADPNGDSPA NVDAAKEWRE DRNGEFKRKV ARCVRKSQET AFE
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
UBE2G protein solution (0.5 mg/ml) containing Phosphate buffered saline, (pH 7.4) 30% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers