missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human UbcH7/UBE2L3 His (N-Term) Protein

Product Code. 18408251 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
1mg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408251 500 μg 500µg
18450162 1mg 1mg
18454782 10 μg 10µg
18412212 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18408251 Supplier Novus Biologicals™ Supplier No. NBP235110500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) PAGE
Formulation A 0.2 um filtered concentrated solution in 50 mM HEPES, pH 7.0, with 125 mM NaCl, 10 % Glycerol, 1 mM DTT.
Gene ID (Entrez) 7332
Molecular Weight (g/mol) M.W. (Theoretical): 18.9 kDa
Quantity 500 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/μg of UbcH7/UBE2L3 as determined by LAL method.
Gene Alias E2-F1, EC 6.3.2.19, L-UBC, UBCE7, UbcH7, UBCH7UbcM4, Ubiquitin carrier protein L3, ubiquitin-conjugating enzyme E2 L3, Ubiquitin-conjugating enzyme E2-F1, ubiquitin-conjugating enzyme E2L 3, ubiquitin-conjugating enzyme UBCH7, Ubiquitin-protein ligase L3
Gene Symbol UBE2L3
Research Category Biologically Active Proteins
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein UbcH7/UBE2L3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.