missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human UbcH5c/UBE2D3 His (N-Term) Protein

Product Code. 18433302 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433302 500 μg 500µg
18470242 1 mg 1mg
18488891 10 μg 10µg
18453032 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18433302 Supplier Novus Biologicals™ Supplier No. NBP235092500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P61077
For Use With (Application) PAGE
Formulation A 0.2 um filtered concentrated solution in 50 mM HEPES, pH 6.5, with 125 mM NaCl, 10 % Glycerol, 1 mM DTT.
Gene ID (Entrez) 7323
Molecular Weight (g/mol) 17.7 kDa
Quantity 500 μg
Storage Requirements Store at -20C. Avoid freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of UbcH5c/UBE2D3, His as determined by LAL method.
Gene Alias E2(17)KB 3, E2(17)KB3, EC 6.3.2.19, MGC43926, MGC5416, UBC4/5, UBC5C, UBCH5C, Ubiquitin carrier protein D3, ubiquitin-conjugating enzyme E2 D3, Ubiquitin-conjugating enzyme E2(17)KB 3, Ubiquitin-conjugating enzyme E2-17 kDa 3, ubiquitin-conjugating enzyme E2D 3 (homologous to yeast UBC4/5), ubiquitin-conjugating enzyme E2D 3 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase D3
Gene Symbol UBE2D3
Reconstitution Please contact technical support for detailed reconstitution instructions.
Research Category Biologically Active Proteins
Purity or Quality Grade >95% by SDS-PAGE and HPLC
Protein UbcH5c/UBE2D3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.