missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human u-Plasminogen Activator/Urokinase
Description
The Recombinant Human u-Plasminogen Activator/Urokinase is derived from NS0. The Recombinant Human u-Plasminogen Activator/Urokinase has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in HEPES, NaCl and CaCl 2 . |
| Gene ID (Entrez) | 5328 |
| Molecular Weight (g/mol) | 18 kDa (A chain), 30 kDa (B chain) |
| Name | u-Plasminogen Activator/Urokinase |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived human u-Plasminogen Activator (uPA)/Urokinase protein Met1-Leu431 with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Gene Alias | ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, PLAU, u-PA, UPA, uPlasminogen Activator, U-plasminogen activator, urinary, Urokinase, urokinase-type plasminogen activator |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?