missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tyrosine-protein kinase ABL1 Protein

A DNA sequence encoding the Homo sapiens (Human) Tyrosine-protein kinase ABL1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8475-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16034821

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_005148.2
Tyrosine-protein kinase ABL1 Protein
Research Use Only
ABL
Recombinant Protein
E. coli
ICLKLVGCKS KKGLSSSSSC YLEEALQRPV ASDFEPQGLS EAARWNSKEN LLAGPSENDP NLFVALYDFV ASGDNTLSIT KGEKLRVLGY NHNGEWCEAQ TKNGQGWVPS NYITPVNSLE KHSWYHGPVS RNAAEYLLSS GINGSFLVRE SESSPGQRSI SLRYEGRVYH YRINTASDGK LYVSSESRFN T
Greater than 90% as determined by SDS-PAGE.
25
200 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers