missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TWEAK R/TNFRSF12 protein is fully biologically active when compared to standard. The ED50 as determined by inhibiting TWEAK-dependent proliferation of human umbilical vein endothelial cells (HUVEC) is less than 30 ng/mL, corresponding to a specific activity of >3.3 x 104 IU/mg, in the presence of 15 ng/mL of rHuTWEAK
Specifications
Specifications
| Accession Number | Q9NP84, Q9NP84, Q9NP84 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 51330 |
| Molecular Weight (g/mol) | 5.6 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of TWEAK R/TNFRSF12 as determined by LAL method. |
| Gene Alias | CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, FN14tumor necrosis factor receptor superfamily member 12A, tumor necrosis factor receptor superfamily, member 12A, TWEAK Receptor/TNFRSF12A, TweakR, tweak-receptor, type I transmembrane protein Fn14 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?