missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tubulin-specific chaperone C Protein
A DNA sequence encoding the Homo sapiens (Human) Tubulin-specific chaperone C, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_003183.1 |
---|---|
Gene ID (Entrez) | 6903 |
Name | Tubulin-specific chaperone C Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16075650
|
enQuireBio™
QP6763-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16095650
|
enQuireBio™
QP6763-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16085650
|
enQuireBio™
QP6763-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005660
|
enQuireBio™
QP6763-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015660
|
enQuireBio™
QP6763-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16025660
|
enQuireBio™
QP6763-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_003183.1 | |
Tubulin-specific chaperone C Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
6903 | |
Research Use Only | |
TBCC | |
Recombinant Protein | |
E. coli | |
ESVSCSAAAV RTGDMESQRD LSLVPERLQR REQERQLEVE RRKQKRQNQE VEKENSHFFV ATFVRERAAV EELLERAESV ERLEEAASRL QGLQKLINDS VFFLAAYDLR QGQEALARLQ AALAERRRGL QPKKRFAFKT RGKDAASSTK VDAAPGIPPA VESIQDSPLP KKAEGDLGPS WVCGFSNLES QVLEKRASEL HQRDVLLTEL SNCTVRLYGN PNTLRLTKAH SCKLLCGPVS TSVFLEDCSD CVLAVACQQL RIHSTKDTRI FLQVTSRAIV EDCSGIQFAP YTWSYPEIDK DFESSGLDRS KNNWNDVDDF NWLARDMASP NWSILPEEER NIQW | |
Greater than 90% as determined by SDS-PAGE. |