missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&DSystems™ Recombinant Human TSPAN14-LEL Fc Chimera Protein, CF
Click to view available options
:
100 μg
Description
Bioassay data are not available.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 81619 |
| Molecular Weight (g/mol) | M.W.-observed: 43-54 kDa, under reducing conditions, M.W.-theroretical: 40 kDa |
| Quantity | 100 μg |
| Source | Human embryonic kidney cell, HEK293-derived human TSPAN14 protein MD Human IgG1 (Pro100-Lys330) IEGR Human TSPAN14-LEL (LEU114-Asn232) (N-terminus) Accession # Q8NG11.1 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | DC-TM4F2, tetraspanin 14, Tetraspanin-14, TM4SF14, transmembrane 4 superfamily member 14 |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction