missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human TSH alpha/beta Heterodimer Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 4610-TH-010/CF

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18391418

  • £399.56 / 10µg
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human TSH alpha/beta Heterodimer Protein is derived from CHO. The Recombinant Human TSH alpha/beta Heterodimer Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
10.2 kDa (TSH alpha), 13.5 kDa (TSH beta)
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
7252
TSH alpha/beta
Chinese Hamster Ovary cell line,CHO-derived human TSH alpha/beta Heterodimer protein, (N-terminus)Human TSH alpha (Ala25-Ser116) Accession &Num; P01215 Human TSH beta (Phe21-Val138) Accession &Num; P01222 (C-terminus)
Noncovalently-linked heterodimer
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers