missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TSH alpha/beta Heterodimer Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 4610-TH-010
Additional Details : Weight : 0.00970kg
Description
The Recombinant Human TSH alpha/beta Heterodimer Protein is derived from CHO. The Recombinant Human TSH alpha/beta Heterodimer Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
10.2 kDa (TSH alpha), 13.5 kDa (TSH beta) | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
Reconstitute at 100μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin. |
7252 | |
TSH alpha/beta | |
Chinese Hamster Ovary cell line,CHO-derived human TSH alpha/beta Heterodimer protein, (N-terminus)Human TSH alpha (Ala25-Ser116) Accession &Num; P01215 Human TSH beta (Phe21-Val138) Accession &Num; P01222 (C-terminus) | |
Noncovalently-linked heterodimer | |
Recombinant | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |