missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Trypsin-2 Protein
Click to view available options
Quantity:
1 mg
5 mg
50 mg
Unit Size:
1mg
50mg
5mg
Specifications
Specifications
| Name | Trypsin-2 Protein |
| Quantity | 50 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | MNLLLILTFV AAAVAAPFDD DDKIVGGYIC EENSVPYQVS LNSGYHFCGG SLISEQWVVS AGHCYKSRIQ VRLGEHNIEV LEGNEQFINA AKIIRHPKYN SRTLDNDILL IKLSSPAVIN SRVSAISLPT APPAAGTESL ISGWGNTLSS GADYPDELQC LDAPVLSQAE CEASYPGKIT NNMFCVGFLE GGKDSCQGDS GGPVVSNGEL QGIVSWGYGC AQKNRPGVYT KVYNYVDWIK DTIAANS |
| Buffer | The protein was lyophilized without any additives. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction