missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Trypsin-1 Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | Trypsin-1 Protein |
| Quantity | 20 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | 1 unit hydrolyzes 1 micromole of Na-Benzoyl-D,L-arginine 4-nitroanilide (BANI) per minute at 37C, pH 8.0, in the presence of 10mM calcium. The specific activity is >1,000 Units per mg protein as measured under the described conditions. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | Human |
| Protein Tag | Untagged |
| Sequence | IVGGYNCEEN SVPYQVSLNS GYHFCGGSLI NEQWVVSAGH CYKSRIQVRL GEHNIEVLEG NEQFINAAKI IRHPQYDRKT LNNDIMLIKL SSRAVINARV STISLPTAPP ATGTKCLISG WGNTASSGAD YPDELQCLDA PVLSQAKCEA SYPGKITSNM FCVGFLEGGK DSCQGDSGGP VVCNGQLQGV VSWGDGCAQK NKPGVYTKVY NYVKWIKNTI AANS |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion