missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TREX2 Protein
A cDNA sequence encoding the TREX2 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | TREX2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944789
|
enQuireBio™
QP13813-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934789
|
enQuireBio™
QP13813-25UG |
25 μg |
£155.00
25µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924789
|
enQuireBio™
QP13813-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TREX2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TREX2 protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 5mM DTT and 30% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSEAPRAETF VFLDLEATGL PSVEPEIAEL SLFAVHRSSL ENPEHDESGA LVLPRVLDKL TLCMCPERPF TAKASEITGL SSEGLARCRK AGFDGAVVRT LQAFLSRQAG PICLVAHNGF DYDFPLLCAE LRRLGARLPR DTVCLDTLPA LRGLDRAHSH GTRARGRQGY SLGSLFHRYF RAEPSAAHSA EGDVHTLLLI FLHRAAELLA WADEQARGWA HIEPMYLPPD DPSLEA | |
Greater than 95% as determined by SDS-PAGE. |