missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human TRAP alpha His Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.02 mg
0.1 mg
Unit Size:
0.02mg
0.1mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 22-207 of Human TRAP alpha The Recombinant Human TRAP alpha Protein is derived from E. coli. The Recombinant Human TRAP alpha Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 0.25mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl(pH8.0) containing 20% glycerol, 0.1M NaCl, 1mM DTT |
| Gene ID (Entrez) | 6745 |
| Molecular Weight (g/mol) | TMW: 23.1kDa |
| Quantity | 0.02 mg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | DKFZp781N23103, FLJ14232, FLJ22100, FLJ23034, FLJ78242, FLJ93042, signal sequence receptor, alpha, SSR alpha subunit, SSR-alpha, translocon-associated protein alpha subunit, translocon-associated protein subunit alpha, TRAP alpha, TRAP-alpha, TRAPASignal sequence receptor subunit alpha |
| Common Name | TRAP alpha |
| Conjugate | Unconjugated |
| Show More |
For research use only and is not approved for use in humans or in clinical diagnosis
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction