missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transglutaminase II Protein

Product Code. p-7153623
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16065820

Brand: enQuireBio™ QP6784ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Transglutaminase II, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001310245.1
Gene ID (Entrez) 7052
Name Transglutaminase II Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol TGM2
Protein Length Full Length of Isoform 2
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence ELVLERCDLE LETNGRDHHT ADLCREKLVV RRGQPFWLTL HFEGRNYEAS VDSLTFSVVT GPAPSQEAGT KARFPLRDAV EEGDWTATVV DQQDCTLSLQ LTTPANAPIG LYRLSLEAST GYQGSSFVLG HFILLFNAWC PADAVYLDSE EERQEYVLTQ QGFIYQGSAK FIKNIPWNFG QFEDGILDIC LILLDVNPKF LKNAGRDCSR RSSPVYVGRV VSGMVNCNDD QGVLLGRWDN NYGDGVSPMS WIGSVDILRR WKNHGCQRVK YGQCWVFAAV ACTVLRCLGI PTRVVTNYNS AHDQNSNLLI EYFRNEFGEI QGDKSEMIWN FHCWVESWMT RPDLQPGYEG WQALDPTPQE KSEGTYCCGP VPVRAIKEGD LSTKYDAPFV FAEVNADVVD WIQQDDGSVH KSINRSLIVG LKISTKSVGR DEREDITHTY KYPEGSSEER EAFTRANHLN KLAEKEETGM AMRIRVGQSM NMGSDFDVFA HITNNTAEEY VCRLLLCART VSYNGILGPE CGTKYLLNLN LEPFSGKALC S
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.