missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Transglutaminase 2/TGM2 Protein
Description
The Recombinant Human Transglutaminase 2/TGM2 Protein is derived from Sf 21 (baculovirus). The Recombinant Human Transglutaminase 2/TGM2 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl, Glycerol and DTT. |
| Gene ID (Entrez) | 7052 |
| Molecular Weight (g/mol) | 78 kDa |
| Name | Transglutaminase 2/TGM2 |
| Quantity | 50 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Transglutaminase 2/TGM2 protein Ala2-Ala687 (Asn533Thr and Leu655Val),with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?