missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transcriptional enhancer factor TEF-5 Protein

Product Code. 16002271
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16002271 100 μg 100µg
16012271 10 μg 10µg
16052271 50 μg 50µg
16032271 200 μg 200µg
16042271 500 μg 500µg
16022271 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16002271 Supplier enQuireBio™ Supplier No. QP8032ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Transcriptional enhancer factor TEF-5, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_003205.2
Gene ID (Entrez) 7005
Name Transcriptional enhancer factor TEF-5 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol TEAD3
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MNLDQVSKDK ALQSMASMSS AQIVSASVLQ NKFSPPSPLP QAVFSTSSRF WSSPPLLGQQ PGPSQDIKPF AQPAYPIQPP LPPTLSSYEP LAPLPSAAAS VPVWQDRTIA SSRLRLLEYS AFMEVQRDPD TYSKHLFVHI GQTNPAFSDP PLEAVDVRQI YDKFPEKKGG LKELYEKGPP NAFFLVKFWA DLNSTIQEGP GAFYGVSSQY SSADSMTISV STKVCSFGKQ VVEKVETEYA RLENGRFVYR IHRSPMCEYM INFIHKLKHL PEKYMMNSVL ENFTILQVVT SRDSQETLLV IAFVFEVSTS EHGAQHHVYK LVKD
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.