missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcription factor SOX-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcription factor SOX-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6718-ec-50ug
Additional Details : Weight : 0.01000kg
Spécification
NP_003097.1 | |
Transcription factor SOX-2 Protein | |
Research Use Only | |
SOX2 | |
Recombinant Protein | |
E. coli | |
MYNMMETELK PPGPQQTSGG GGGNSTAAAA GGNQKNSPDR VKRPMNAFMV WSRGQRRKMA QENPKMHNSE ISKRLGAEWK LLSETEKRPF IDEAKRLRAL HMKEHPDYKY RPRRKTKTLM KKDKYTLPGG LLAPGGNSMA SGVGVGAGLG AGVNQRMDSY AHMNGWSNGS YSMMQDQLGY PQHPGLNAHG AAQMQPMHRY DVSALQYNSM TSSQTYMNGS PTYSMSYSQQ GTPGMALGSM GSVVKSEASS SPPVVTSSSH SRAPCQAGDL RDMISMYLPG AEVPEPAAPS RLHMSQHYQS GPVPGTAING TLPLSHM | |
Greater than 90% as determined by SDS-PAGE. |
6657 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |