missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transcobalamin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Transcobalamin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6769-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015700

  • £1735.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001053.2
Transcobalamin-1 Protein
Research Use Only
TCN1
Recombinant Protein
E. coli
EICEVSEENY IRLKPLLNTM IQSNYNRGTS AVNVVLSLKL VGIQIQTLMQ KMIQQIKYNV KSRLSDVSSG ELALIILALG VCRNAEENLI YDYHLIDKLE NKFQAEIENM EAHNGTPLTN YYQLSLDVLA LCLFNGNYST AEVVNHFTPE NKNYYFGSQF SVDTGAMAVL ALTCVKKSLI NGQIKADEGS LKNISIYTKS LVEKILSEKK ENGLIGNTFS TGEAMQALFV SSDYYNENDW NCQQTLNTVL TEISQGAFSN PNAAAQVLPA LMGKTFLDIN KDSSCVSASG NFNISADEPI TVTPPDSQSY ISVNYSVRIN ETYFTNVTVL NGSVFLSVME KAQKMNDTIF GFTMEERSWG PYITCIQGLC ANNNDRTYWE LLSGGEPLSQ GAGSYVVRNG ENLEVRWSKY
Greater than 90% as determined by SDS-PAGE.
6947
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers