missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human TRANCE/RANK L/TNFSF11 Protein

Product Code. 18117110
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
10µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18117110 Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10µg
18334987 Lyophilized from a 0.2μm filtered solution in PBS. 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18117110 Supplier R&D Systems™ Supplier No. 390TN010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human TRANCE/RANK L/TNFSF11 Protein is derived from NS0. The Recombinant Human TRANCE/RANK L/TNFSF11 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 8600
Molecular Weight (g/mol) 23 kDa
Name TRANCE/TNFSF11/RANK L
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived human TRANCE/TNFSF11/RANK L protein, (N-terminus) MHHHHHHHHHH GGGSGGGSGGGS IEGR Human TRANCE (Gly64-Asp245) Accession &Num; AAC51762 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.