missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TRAILR1/TNFRSF10A Fc Chimera Protein
Description
The Recombinant Human TRAIL R1/TNFRSF10A Fc Chimera Protein is derived from Sf 21 (baculovirus). The Recombinant Human TRAIL R1/TNFRSF10A Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 8797 |
| Molecular Weight (g/mol) | 43 kDa (monomer) |
| Name | TRAILR1/TNFRSF10A |
| Quantity | 100 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human TRAIL R1/TNFRSF10A protein, (N-terminus) Human TRAIL R1 (Ala109-Asn239) & (Thr110-Asn239) Accession &Num; AAC51226 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Gene Alias | APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TNFRSF10A, TRAIL R1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?