missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human TRAIL R2/TNFRSF10B Isoform 1 Protein

Product Code. 18202964 Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18202964

Brand: Novus Biologicals™ NBP26198310UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page, Bioactivity

A Biologically active recombinant protein with a C-Terminal hIgG-His-tag and corresponding to the amino acids 56-210 of Human TRAIL R2/TNFRSF10B The Recombinant Human TRAILR2/TNFRSF10B Isoform 1 Protein is derived from Baculovirus. The Recombinant Human TRAILR2/TNFRSF10B Isoform 1 Protein has been validated for the following applications: SDS-Page, Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/ml
For Use With (Application) Bioactivity, SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4) containing 10% glycerol.
Gene ID (Entrez) 8795
Molecular Weight (g/mol) 43.9 kDa
Quantity 10 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1μg of protein (determined by LAL method)
Gene Symbol TNFRSF10B
Cross Reactivity TRAIL R2/TNFRSF10B
Species Human
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.