missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TRACP/PAP/ACP5 Protein
Description
Measured by its ability to cleave a substrate, p-Nitrophenyl phosphate (pNPP). The specific activity is >65,000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | NP_001602 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 54 |
| Molecular Weight (g/mol) | M.W. (Observed): 36-40 kDa, reducing conditions; M.W. (theoretical): 34 kDa |
| Name | TRACP/PAP/ACP5 |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line, NS0-derived human TRACP/PAP/ACP5 protein Ala22-Pro320, with a C-terminal 6-His tag |
| Storage Requirements | Do not freeze. 3 months from date of receipt, 2°C to 8°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?