missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TMED10 Protein
A cDNA sequence encoding the TMED10 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13761-10ug
Additional Details : Weight : 0.01000kg
Specifications
10972 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSISFHLPI NSRKCLREEI HKDLLVTGAY EISDQSGGAG GLRSHLKITD SAGHILYSKE DATKGKFAFT TEDYDMFEVC FESKGTGRIP DQLVILDMKH GVEAKNYEEI AKVEKLKPLE VELRRLEDLS ESIVNDFAYM KKREEEMRDT NESTNTR | |
Greater than 85% as determined by SDS-PAGE. |
TMED10 Protein | |
Research Use Only | |
TMED10 | |
Human | |
His | |
The TMED10 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 30% glycerol. |