missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TL1A/TNFSF15 protein is fully biologically active when compared to standard. The ED50 as determined by its ability to induce apoptosis using human TF-1 cells is less than 20 ng/mL, corresponding to a specific activity of >5.0 x 10^4 IU/mg
Specifications
Specifications
| Accession Number | O95150, O95150, O95150 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4, with 0.02% Tween-20. |
| Gene ID (Entrez) | 9966 |
| Molecular Weight (g/mol) | 20.5 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of TL1A/TNFSF15 as determined by LAL method. |
| Gene Alias | MGC129934, MGC129935, TL1A, TL-1A, TL-1A/TNFSF15, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI192A, VEGITNF ligand-related molecule 1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?