missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TKT Protein
A cDNA sequence encoding the TKT was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13758-10ug
Additional Details : Weight : 0.01000kg
Specifications
TKT Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MESYHKPDQQ KLQALKDTAN RLRISSIQAT TAAGSGHPTS CCSAAEIMAV LFFHTMRYKS QDPRNPHNDR FVLSKGHAAP ILYAVWAEAG FLAEAELLNL RKISSDLDGH PVPKQAFTDV ATGSLGQGLG AACGMAYTGK YFDKASYRVY CLLGDGELSE GSVWEAMAFA SIYKLDNLVA ILDINRLGQS DPAPLQHQMD IYQKRCEAFG WHAIIVDGHS VEELCKAFGQ AKHQPTAIIA KTFKGRGITG VEDKESWHGK PLPKNMAEQI IQEIYSQIQS KKKILATPPQ EDAPSVDIAN IRMPSLPSYK VGDKIATRKA YGQALAKLGH ASDRIIALDG DTKNSTFSEI FKKEHPDRFI ECYIAEQNMV SIAVGCATRN RTVPFCSTFA AFFTRAFDQI RMAAISESNI NLCGSHCGVS IGEDGPSQMA LEDLAMFRSV PTSTVFYPSD GVATEKAVEL AANTKGICFI RTSRPENAII YNNNEDFQVG QAKVVLKSKD DQVTVIGAGV TLHEALAAAE LLKKEKINIR VLDPFTIKPL DRKLILDSAR ATKGRILTVE DHYYEGGIGE AVSSAVVGEP GITVTHLAVN RVPRSGKPAE LLKMFGIDRD AIAQAVRGLI TKA | |
Greater than 85% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TKT solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM Nacl, 1mM DTT and 10% glycerol. |