missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TIMM23 Protein

A DNA sequence encoding the Homo sapiens (Human) TIMM23, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6793-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16075890

  • £384.00 / 100µg
Estimated Shipment: 16-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006318.1
TIMM23 Protein
Research Use Only
TIMM23
Recombinant Protein
E. coli
MEGGGGSGNK TTGGLAGFFG AGGAGYSHAD LAGVPLTGMN PLSPYLNVDP RYLVQDTDEF ILPTGANKTR GRFELAFFTI GGCCMTGAAF GAMNGLRLGL KETQNMAWSK PRNVQILNMV TRQGALWANT LGSLALLYSA FGVIIEKTRG AEDDLNTVAA GTMTGMLYKC TGGLRGIARG GLTGLTLTSL YALYNNWEHM KGSLLQQSL
Greater than 80% as determined by SDS-PAGE.
100287932
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers