missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TIGAR protein is fully biologically active when compared to standard. The biological activity determined by its ability to protect U2OS cells from apoptosis induced by hydrogen peroxide is in a concentration range of 0.1-5.0 μg/mL, after pretreating with rHuTIGAR-TAT for 4 hours.
Specifications
Specifications
| Accession Number | Q9NQ88 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 30 % Acetonitrile, 0.1% TFA. |
| Gene ID (Entrez) | 57103 |
| Molecular Weight (g/mol) | 31.7 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of TIGAR/C12orf5 as determined by LAL method. |
| Gene Alias | chromosome 12 open reading frame 5, EC 3.1.3.46, probable fructose-2,6-bisphosphatase TIGAR, TIGARTP53-induced glycolysis and apoptosis regulator, transactivated by NS3TP2 protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?