missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Thymosin beta 4 protein is fully biologically active when compared to standard. The biological activity determined by its ability to produce a protective effect against hydrogen peroxide in primary lung fibroblasts is in a concentration range of 0.5 - 10 μg/mL.
Specifications
Specifications
| Accession Number | P62328 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4. |
| Gene ID (Entrez) | 7114 |
| Molecular Weight (g/mol) | 4.9 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of Thymosin beta 4 as determined by LAL method. |
| Gene Alias | PTMB4, T beta-4, TB4, TB4Xbeta 4, X chromosome, THYB4, thymosin beta 4, X-linked, thymosin beta-4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?