missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Thrombospondin-1 Protein

Product Code. 16025341
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16025341 100 μg 100µg
16035341 10 μg 10µg
16075341 50 μg 50µg
16055341 200 μg 200µg
16065341 500 μg 500µg
16045341 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16025341 Supplier enQuireBio™ Supplier No. QP8570ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Thrombospondin-1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_003237.2
Gene ID (Entrez) 7057
Name Thrombospondin-1 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol THBS1
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence NRIPESGGDN SVFDIFELTG AARKGSGRRL VKGPDPSSPA FRIEDANLIP PVPDDKFQDL VDAVRAEKGF LLLASLRQMK KTRGTLLALE RKDHSGQVFS VVSNGKAGTL DLSLTVQGKQ HVVSVEEALL ATGQWKSITL FVQEDRAQLY IDCEKMENAE LDVPIQSVFT RDLASIARLR IAKGGVNDNF QGVLQNVRFV FGTTPEDILR NKGCSSSTSV LLTLDNNVVN GSSPAIRTNY IGHKTKDLQA ICGISCDELS SMVLELRGLR TIVTTLQDSI RKVTEENKEL ANELRRPPLC YHNGVQYRNN EEWTVDSCTE CHCQNSVTIC KK
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.