missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Thrombopoietin (NS0-expressed) Protein

Product Code. 18706886
Click to view available options
Quantity:
10 μg
100 μg
Unit Size:
100µg
10µg
This item is not returnable. View return policy

Product Code. 18706886

Brand: R&D Systems™ 288TPN010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A new economical version of TPO made in E. coli is now available.

Measured in a cell proliferation assay using MO7e human megakaryocytic leukemic cells. Avanzi, G. et al. (1988) Br. J. Haematol. 69:359. The ED50 for this effect is 5-15 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P40225
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 7066
Molecular Weight (g/mol) M.W. (Observed): 80-90 kDa, reducing conditions; M.W. (theoretical): 35 kDa
Name Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived human Thrombopoietin/Tpo protein Ser22-Gly353
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol THPO
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.1 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Thrombopoietin/THPO
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.