missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Thrombin Protein

Product Code. 15985448
Change view
Click to view available options
Quantity:
1 mg
5 mg
500 μg
Unit Size:
1mg
500µg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15985448 500 μg 500µg
15975448 1 mg 1mg
15995448 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15985448 Supplier enQuireBio™ Supplier No. QP10880500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Thrombin was constructed and used to recombinantly synthesize the protein.

Specifications

Name Thrombin Protein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity The specific activity was found to be 3005 NIH Units/mg.
Product Type Recombinant Protein
Cross Reactivity Human
Species Human
Protein Tag Untagged
Buffer The Factor-IIa contains 50mM sodium Citrate, 0.2M NaCl and 0.1% PEG-8000 pH 6.5.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.