missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human THOC7 Protein

A cDNA sequence encoding the THOC7 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13743-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944559

  • £5730.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

THOC7 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMGAVTDD EVIRKRLLID GDGAGDDRRI NLLVKSFIKW CNSGSQEEGY SQYQRMLSTL SQCEFSMGKT LLVYDMNLRE MENYEKIYKE IECSIAGAHE KIAECKKQIL QAKRIRKNRQ EYDALAKVIQ HHPDRHETLK ELEALGKELE HLSHIKESVE DKLELRRKQF HVLLSTIHEL QQTLENDEKL SEVEEAQEAS METDPKP
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
THOC7 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4) and 50% glycerol, 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers