missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human THOC7 Protein
A cDNA sequence encoding the THOC7 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13743-10ug
Additional Details : Weight : 0.01000kg
Specifications
THOC7 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGAVTDD EVIRKRLLID GDGAGDDRRI NLLVKSFIKW CNSGSQEEGY SQYQRMLSTL SQCEFSMGKT LLVYDMNLRE MENYEKIYKE IECSIAGAHE KIAECKKQIL QAKRIRKNRQ EYDALAKVIQ HHPDRHETLK ELEALGKELE HLSHIKESVE DKLELRRKQF HVLLSTIHEL QQTLENDEKL SEVEEAQEAS METDPKP | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
THOC7 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4) and 50% glycerol, 1mM DTT. |