missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TGFBR3 Protein

Product Code. 15924068
Click to view available options
Quantity:
10 μg
2 μg
5 μg
Unit Size:
10µg
2µg
5µg
This item is not returnable. View return policy

Product Code. 15924068

Brand: enQuireBio™ QP105015ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the TGFBR3 was constructed and used to recombinantly synthesize the protein.

Specifications

Name TGFBR3 Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence PYSNPDRMSH YTIIENICPK DESVKFYSPK RVHFPIPQAD MDKKRFSFVF KPVFNTSLLF LQCELTLCTK MEKHPQKLPK CVPPDEACTS LDASIIWAMM QNKKTFTKPL AVIHHEAESK EKGPSMKEPN PISPPIFHGL DTLTVMGIAF AAFVIGALLT GALWYIYSHT GETAGRQQVP TSPPASENSS AAHSIGSTQS TPCSSSS
Buffer 100microL purified human TGF Beta Receptor 3 protein at 100microg/ml in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.