missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TGFB3 Protein

A DNA sequence encoding the Homo sapiens (Human) TGFB3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6783-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015820

  • £1735.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001316867.1
TGFB3 Protein
Research Use Only
TGFB3
Recombinant Protein
E. coli
LSTCTTLDFG HIKKKRVEAI RGQILSKLRL TSPPEPTVMT HVPYQVLALY NSTRELLEEM HGEREEGCTQ ENTESEYYAK EIHKFDMIQG LAEHNELAVC PKGITSKVFR FNVSSVEKNR TNLFRAEFRV LRVPNPSSKR NEQRIELFQI LRPDEHIAKQ RYIGGKNLPT RGTAEWLSFD VTDTVREWLL RRESNLGLEI SIHCPCHTFQ PNGDILENIH EVMEIKFKGV DNEDDHGRGD LGRLKKQKDH HNPHLILMMI PPHRLDNPGQ GGQRKKR
Greater than 90% as determined by SDS-PAGE.
7043
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers