missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TGF-beta RIII Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£530.22
Specifications
Gene ID (Entrez) | 7049 |
---|---|
Molecular Weight (g/mol) | 84 kDa |
Name | TGF-beta RIII |
Quantity | 100μg |
Source | Mouse myeloma cell line,NS0-derived human TGF-beta RIII protein Gly21-Asp781 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18128020
|
R&D Systems™
242-R3-100/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£530.22
100µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18106310
|
R&D Systems™
242-R3-100 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£530.22
100µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human TGF-beta RIII Protein is derived from NS0. The Recombinant Human TGF-beta RIII Protein has been validated for the following applications: Bioactivity.Specifications
7049 | |
TGF-beta RIII | |
Mouse myeloma cell line,NS0-derived human TGF-beta RIII protein Gly21-Asp781 | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
84 kDa | |
100μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |